ENA360886891 Enamine

4-butyl-N-methylaniline hydrochloride

  • MDL number MFCD21602734
  • Cas Number : 1375472-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98798
Availability details for
Quantity
  • Availability
  • Add to Cart