ENA360886892 Enamine

1-(2-chloro-6-fluorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD21602735
  • Cas Number : 1375474-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196...198
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98801
Availability details for
Quantity
  • Availability
  • Add to Cart