ENA360886894 Enamine

3-(pentan-2-yloxy)aniline hydrochloride

  • MDL number MFCD21602737
  • Cas Number : 1375474-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98807
Availability details for
Quantity
  • Availability
  • Add to Cart