ENA360886902 Enamine

1-[2-hydroxy-4-(pyrrolidin-1-yl)phenyl]ethan-1-one

  • MDL number MFCD21602745
  • Cas Number : 1375471-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-98994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98994
Availability details for
Quantity
  • Availability
  • Add to Cart