ENA360886905 Enamine

1-methyl-2-phenyl-1H,6H,7H-pyrrolo[2,3-c]pyridin-7-one

  • MDL number MFCD21602748
  • Cas Number : 1375473-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-99000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99000
Availability details for
Quantity
  • Availability
  • Add to Cart