ENA360886906 Enamine

2-[4-(aminomethyl)phenoxy]-1-(morpholin-4-yl)ethan-1-one hydrochloride

  • MDL number MFCD21602749
  • Cas Number : 1375471-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99005
Availability details for
Quantity
  • Availability
  • Add to Cart