ENA360886907 Enamine

ethyl 2-amino-3-cyclopropylpropanoate hydrochloride

  • MDL number MFCD21602750
  • Cas Number : 1375471-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99021
Availability details for
Quantity
  • Availability
  • Add to Cart