ENA360886909 Enamine

1-[(1-methyl-1H-imidazol-2-yl)methyl]piperidine-4-carboxylic acid dihydrochloride

  • MDL number MFCD21602752
  • Cas Number : 1375471-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99031
Availability details for
Quantity
  • Availability
  • Add to Cart