ENA360886925 Enamine

2-methylthiolan-3-ol

  • MDL number MFCD21602771
  • Cas Number : 149834-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99127
Availability details for
Quantity
  • Availability
  • Add to Cart