ENA360886945 Enamine

7-methyl-2,3-dihydro-1H-indole hydrochloride

  • MDL number MFCD21602791
  • Cas Number : 1375472-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99212

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99212
Availability details for
Quantity
  • Availability
  • Add to Cart