ENA360886952 Enamine

5-(dimethylamino)-2-nitrophenol

  • MDL number MFCD00047680
  • Cas Number : 14703-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99266
Availability details for
Quantity
  • Availability
  • Add to Cart