ENA360886957 Enamine

4-hydroxypiperidine-4-carbonitrile hydrochloride

  • MDL number MFCD21602802
  • Cas Number : 1375472-85-7
  • Molecular Weight : 162.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-99300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99300
Availability details for
Quantity
  • Availability
  • Add to Cart