ENA360886959 Enamine

2-{[(2-ethyl-1,3-thiazol-5-yl)methyl]amino}acetic acid dihydrochloride

  • MDL number MFCD21602804
  • Cas Number : 1375472-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99320
Availability details for
Quantity
  • Availability
  • Add to Cart