ENA360886961 Enamine

4-(propan-2-yl)piperidin-4-ol hydrochloride

  • MDL number MFCD21602808
  • Cas Number : 1246632-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99380
Availability details for
Quantity
  • Availability
  • Add to Cart