ENA360886962 Enamine

4,4,4-trifluoro-3-hydroxy-3-methylbutanoic acid

  • MDL number MFCD00039531
  • Cas Number : 338-03-4
  • Molecular Weight : 172.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39-41
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14885

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-14885
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart