ENA360886965 Enamine

5-iodo-2,2-dimethylpentanenitrile

  • MDL number MFCD21132253
  • Cas Number : 56475-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84936

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84936
Availability details for
Quantity
  • Availability
  • Add to Cart