ENA360886978 Enamine

3-(4-methoxyphenyl)butan-2-one

  • MDL number MFCD21602558

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97026

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97026
Availability details for
Quantity
  • Availability
  • Add to Cart