ENA360887036 Enamine

4-(2-methyl-1,3-oxazol-4-yl)benzene-1-sulfonamide

  • MDL number MFCD11650733
  • Cas Number : 1099660-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245...247
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97191

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97191
Availability details for
Quantity
  • Availability
  • Add to Cart