ENA360887042 Enamine

1-(4-nitrophenyl)-1,2,3,6-tetrahydropyridazine-3,6-dione

  • MDL number MFCD21602606
  • Cas Number : 22202-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97263
Availability details for
Quantity
  • Availability
  • Add to Cart