ENA360887043 Enamine

4-(1,3-dithiolan-2-yl)-N-(3-hydroxypyridin-2-yl)benzamide

  • MDL number MFCD11774214
  • Cas Number : 1060968-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91308
Availability details for
Quantity
  • Availability
  • Add to Cart