ENA360887068 Enamine

2-amino-2-methyl-4-(propan-2-yloxy)butanenitrile

  • MDL number MFCD16135163
  • Cas Number : 1247180-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-93311
Availability details for
Quantity
  • Availability
  • Add to Cart