ENA360887074 Enamine

4-(thiophen-2-yl)-1H,4H,5H,6H,7H-imidazo[4,5-c]pyridine-6-carboxylic acid

  • MDL number MFCD04448841
  • Cas Number : 1030285-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-93834

Safety Information

msds https://www.enaminestore.com/catalog/EN300-93834
Availability details for
Quantity
  • Availability
  • Add to Cart