ENA360887078 Enamine

4-trifluoromethanesulfonylbenzene-1-thiol

  • MDL number MFCD12174234
  • Cas Number : 63647-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97617
Availability details for
Quantity
  • Availability
  • Add to Cart