ENA360887085 Enamine

2-methoxy-1-(5-methylfuran-2-yl)ethan-1-amine

  • MDL number MFCD09734891
  • Cas Number : 953889-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97640
Availability details for
Quantity
  • Availability
  • Add to Cart