ENA360887093 Enamine

3-fluoro-N-[(1H-imidazol-2-yl)methyl]aniline

  • MDL number MFCD14693417
  • Cas Number : 1020670-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-97703
Availability details for
Quantity
  • Availability
  • Add to Cart