ENA360887096 Enamine

5-(chloromethyl)-1-methyl-1H-imidazole hydrochloride

  • MDL number MFCD08668158
  • Cas Number : 90773-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-97717

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-97717
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart