ENA360887098 Enamine

1-(2-fluorobenzoyl)-1,4-diazepane

  • MDL number MFCD09044069
  • Cas Number : 926213-29-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-31028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31028
Availability details for
Quantity
  • Availability
  • Add to Cart