ENA360887112 Enamine

(1R,3S,4S)-2-azabicyclo[2.2.1]heptane-3-carboxylic acid hydrochloride

  • MDL number MFCD08457558
  • Cas Number : 99940-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33707
Availability details for
Quantity
  • Availability
  • Add to Cart