ENA360887126 Enamine

3-(methylsulfonyl)aniline hydrochloride

  • MDL number MFCD00216487
  • Cas Number : 80213-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34402

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-34402
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart