ENA360887127 Enamine

2-[4-(aminomethyl)oxan-4-yl]ethan-1-ol

  • MDL number MFCD21602468
  • Cas Number : 1375163-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95375
Availability details for
Quantity
  • Availability
  • Add to Cart