ENA360887176 Enamine

N-(3-acetylphenyl)-4-ethoxypiperidine-1-carboxamide

  • MDL number MFCD21602683
  • Cas Number : 1304896-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98165
Availability details for
Quantity
  • Availability
  • Add to Cart