ENA360887202 Enamine

3-hydrazinyl-1-methyl-1,2-dihydropyrazin-2-one

  • MDL number MFCD17250001
  • Cas Number : 1342087-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95792
Availability details for
Quantity
  • Availability
  • Add to Cart