ENA360887204 Enamine

N-(4-hydroxycyclohexyl)methanesulfonamide

  • MDL number MFCD12179278
  • Cas Number : 1157109-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98326

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-98326
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart