ENA360887209 Enamine

5-tert-butyl-2-(1-chloroethyl)-1,3-oxazole

  • MDL number MFCD12131302
  • Cas Number : 1133001-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98335
Availability details for
Quantity
  • Availability
  • Add to Cart