ENA360887216 Enamine

[(5-ethyl-1,3-thiazol-2-yl)methyl](methyl)amine

  • MDL number MFCD16834338
  • Cas Number : 1247834-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98373
Availability details for
Quantity
  • Availability
  • Add to Cart