ENA360887219 Enamine

3-chloro-4-(methylsulfanyl)aniline

  • MDL number MFCD00025214
  • Cas Number : 5211-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98446
Availability details for
Quantity
  • Availability
  • Add to Cart