ENA360887221 Enamine

1H-indazole-3,6-diamine

  • MDL number MFCD11707245
  • Cas Number : 871709-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98459
Availability details for
Quantity
  • Availability
  • Add to Cart