ENA360887224 Enamine

N'-hydroxy-2-methoxypropanimidamide

  • Cas Number : 1251430-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98463
Availability details for
Quantity
  • Availability
  • Add to Cart