ENA360887231 Enamine

morpholine-3,5-dione

  • MDL number MFCD01863549

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-145
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53696
Availability details for
Quantity
  • Availability
  • Add to Cart