ENA360887234 Enamine

(1-chloroethyl)cyclopropane

  • MDL number MFCD11186424
  • Cas Number : 10524-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98480
Availability details for
Quantity
  • Availability
  • Add to Cart