ENA360887236 Enamine

3-[(pyridin-4-ylmethyl)amino]propan-1-ol

  • MDL number MFCD00023633
  • Cas Number : 7251-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-98483
Availability details for
Quantity
  • Availability
  • Add to Cart