ENA360887237 Enamine

2,2,2-trifluoroethyl N-[2-methyl-5-(1H-1,2,3,4-tetrazol-1-yl)phenyl]carbamate

  • MDL number MFCD18140508
  • Cas Number : 1241066-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54615
Availability details for
Quantity
  • Availability
  • Add to Cart