ENA360887238 Enamine

1-[2-(2-methylmorpholin-4-yl)pyridin-4-yl]methanamine

  • MDL number MFCD09732848
  • Cas Number : 953901-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98485

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98485
Availability details for
Quantity
  • Availability
  • Add to Cart