ENA360887244 Enamine

4-fluoro-2,3-dihydro-1H-inden-1-amine

  • MDL number MFCD07373911
  • Cas Number : 148960-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58727

Safety Information

msds https://www.enaminestore.com/catalog/EN300-58727
Availability details for
Quantity
  • Availability
  • Add to Cart