ENA360887245 Enamine

1-ethyl-1H-imidazole-4-sulfonyl chloride

  • MDL number MFCD19662430
  • Cas Number : 137049-01-5
  • Molecular Weight : 194.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-98564

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-98564
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart