ENA360887257 Enamine

(2S)-2-amino-3,3-dimethylbutanamide hydrochloride

  • MDL number MFCD21602493
  • Cas Number : 75158-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96001
Availability details for
Quantity
  • Availability
  • Add to Cart