ENA360887259 Enamine

ethyl[(pyrrolidin-3-yl)methyl](2,2,2-trifluoroethyl)amine

  • MDL number MFCD16715014
  • Cas Number : 1250820-34-8
  • Molecular Weight : 210.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-ethyl-2,2,2-trifluoro-N-(3-pyrrolidinylmethyl)ethanamine
Inchi Code 1S/C9H17F3N2/c1-2-14(7-9(10,11)12)6-8-3-4-13-5-8/h8,13H,2-7H2,1H3
Inchi Key YXBLQSNCJULLNT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-98582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98582
Availability details for
Quantity
  • Availability
  • Add to Cart