ENA360887261 Enamine

3-amino-2-[(3,5-dimethoxyphenyl)methyl]propan-1-ol

  • MDL number MFCD11147528
  • Cas Number : 1020943-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-98584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98584
Availability details for
Quantity
  • Availability
  • Add to Cart