ENA360887300 Enamine

(4R)-8-fluoro-3,4-dihydro-2H-1-benzopyran-4-amine hydrochloride

  • MDL number MFCD12912363
  • Cas Number : 730980-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96054
Availability details for
Quantity
  • Availability
  • Add to Cart