ENA360887301 Enamine

3-[(4-methoxypiperidine-1-carbonyl)amino]benzoic acid

  • MDL number MFCD21602718
  • Cas Number : 1308435-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-98732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-98732
Availability details for
Quantity
  • Availability
  • Add to Cart